ContactUsLinkComponent
Molecular model: ten-layer DNA, Molymod®
Molecular model: ten-layer DNA, Molymod®
Catalog # SPIRMKS-122-10
undefined
Molecular model: ten-layer DNA, Molymod®
Catalog # SPIRMKS-122-10
Specifications
  • Description:
    Molecular model of DNA
  • Environmentally Preferable:
  • No. of atoms:
    640
  • Pk:
    1 SET
Specifications
About this item
Molecular model of compact DNA including ten pairs of bases.
  • Height ca. 50 cm
Ordering information: Self-assembly kit incl. 640 atoms and 750 connections on a stand.