Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Kobayashi's Reagent, NaBArF24
CAS Number:79060-88-1
:
Formula:C₃₂H₁₂BF₂₄Na
MDL Number:MFCD00043323
Molecular Weight:886.20 g/mol
:M-Clarity™ quality level = MQ100
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together