4'-Aminobenzanilide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4′-Aminobenzanilide, 4-Aminobenzanilide
CAS Number:17625-83-1
:
Formula:C₁₃H₁₂N₂O
MDL Number:MFCD00035777
Molecular Weight:212.25 g/mol
Melting Pt:127…131 °C
EINECS:241-603-0
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together