Potassium xanthogenate (potassium-O-ethyl dithiocarbonate) for synthesis, Sigma-Aldrich®
: Merck
: Potassium-O-ethyldithiocarbonate, Potassium ethyl xanthate, Ethylxanthic acid potassium salt
:140-89-6
:
:C₃H₅KOS₂
:MFCD00004931
:160.30 g/mol
:M-Clarity™ quality level = MQ200
:205-439-3
:GHS07
:Ambient
- Environmentally Preferable:
- Solubility:1090 g/l (20 °C)
- Melting Point:209 - 214 °C decomposes
- Molar Mass:160.3 g/mol
- Bulk Density:400 - 600 kg/m3
- pH:9 - 10 (100 g/l, H2O, 20 °C)
- Synonyms:Potassium ethylxanthogenate, Potassium xanthogenate
- Hill Formula:C3H5KOS2
Frequently Bought Together