Creatine phosphate disodium salt tetrahydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Sodium creatine phosphate dibasic tetrahydrate, Sodium N-carbamimidoyl-N-methylglycinate dihydrogen phosphate tetrahydrate, Phosphocreatine disodium salt tetrahydrate
CAS Number:71519-72-7
Formula:C₄H₈N₃Na₂O₅P·4H₂O
MDL Number:MFCD00150192
Molecular Weight:327.14 g/mol (255.07 anhy)
Storage Temperature:Refrigerator
EINECS:213-074-6
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together