N-Methyl-N-octanoyl-D-glucamine (MEGA-8)
Supplier: G-Biosciences
Certificates
About this item
Synonyms: N-Octanoyl-N-methyl-D-glucamine, N-Methyl-N-((2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl)octanamide, MEGA-8
CAS Number:85316-98-9
:
Formula:C₁₅H₃₁NO₆
MDL Number:MFCD00134152
Molecular Weight:321.4 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together