Brilliant green ≥90% (dye content), Sigma-Aldrich®
Catalog # B6756-100G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: Ethyl Green, CI 42040, Basic Green 1, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate, Diamond Green, Malachite Green G, Emerald Green, Astradiamant green GX, Solid Green JO, Brilliant Green hydrogen sulphate
:
:482.65 g/mol
EINECS:211-190-1
:GHS07
Storage Temperature:Ambient
CAS Number:633-03-4
Formula:C₂₇H₃₄N₂O₄S
MDL Number:MFCD00011880
Melting Pt:210 °C (dec., lit.)
:M-Clarity™ quality level = MQ200