3,3-Dimethylglutaric anhydride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4,4-Dimethyldihydro-2H-pyran-2,6(3H)-dione
CAS Number:4160-82-1
:
Formula:C₇H₁₀O₃
MDL Number:MFCD00006684
Boiling Pt:181 °C (25 mmHg)
Molecular Weight:142.15 g/mol
Melting Pt:122…126 °C
EINECS:223-998-1
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together