Order Entry
Austria
ContactUsLinkComponent
Triflumuron
Triflumuron
Catalog # EHERC17844300
Supplier:  EHRENSTORFER
CAS Number:  
Triflumuron
Catalog # EHERC17844300
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

 :
 :358.7 g/mol
EINECS:264-980-3
CAS Number:64628-44-0
Formula:C₁₅H₁₀ClF₃N₂O₃
MDL Number:MFCD00072496