2-(Propyloxy)ethanol 98%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Ethylene glycol monopropyl ether, 2-Propoxyethanol, Propyl glycol
CAS Number:2807-30-9
:
Formula:C₅H₁₂O₂
MDL Number:MFCD00072645
Molecular Weight:104.15 g/mol
Melting Pt:–70 °C
Flash Pt:57 °C
Storage Temperature:Ambient
EINECS:220-548-6
UN:1986
ADR:3 III
:GHS06
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4120 to 1.4140 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together