Filter funnels, Gooch, VitraPOR®
Supplier: ROBU GLASFILTER
Certificates
About this item
These laboratory glasswares are made of pure borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Versitile, recyclable materials with high resistent to most chemicals such as solvents, acids and alkalis. With sealed filter disc and standard joint.
Frequently Bought Together