2-(2-Nitroethyl)-1,3-dioxolane for synthesis
Catalog # 8.14136.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 ml
- Seq:2070
- Environmentally Preferable:
Specifications
About this item
Synonyms: 2-(2-Nitroethyl)[1,3]dioxolane
:
:147.13 g/mol
CAS Number:82891-99-4
Formula:C₅H₉NO₄
MDL Number:MFCD00040621
:M-Clarity™ quality level = MQ200