D-(-)-α-Phenylglycine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: D-(-)-2-Phenylglycine, H-D-Phg-OH, (R)-(-)-α-Aminophenylacetic acid
CAS Number:875-74-1
:
Formula:C₈H₉NO₂
MDL Number:MFCD00008061
Molecular Weight:151.16 g/mol
:M-Clarity™ quality level = MQ200
EINECS:212-876-3
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together