Sodium hydroxide, pellets, pure, Supelco®
: Merck
: Caustic soda
:1310-73-2
:
:NaOH
:MFCD00003548
:1390
:323
:2.13 g/cm³ (20 °C)
:40 g/mol
:215-185-5
:1823
:8 II
:14,08627
:M-Clarity™ quality level = MQ200
:Ambient
:01-2119457892-27
:GHS05
- Environmentally Preferable:
- Solubility:1090 g/l (20 °C)
- Melting Point:323 °C
- Molar Mass:40.00 g/mol
- Boiling Point:1390 °C (1013 hPa)
- Vapor Pressure:(20 °C)
- Density:2.13 g/cm3 (20 °C)
- pH:14 (50 g/l, H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Frequently Bought Together