Ethyl 2,4-dichlorobenzoate
Supplier: Thermo Scientific
Certificates
About this item
Synonyms: Ethyl 2,4-dichlorobenzoate
CAS Number:56882-52-1
:
Formula:C₉H₈Cl₂O₂
MDL Number:MFCD00277702
Molecular Weight:219.07 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together