1,3-Dimethoxybenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Resorcinol dimethyl ether
CAS Number:151-10-0
:
Formula:C₈H₁₀O₂
MDL Number:MFCD00008384
Boiling Pt:217…218 °C
Melting Pt:–52 °C
Density:1.065
Molecular Weight:138.17 g/mol
Flash Pt:87 °C (188 °F)
Storage Temperature:Ambient
EINECS:205-783-4
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together