Soxhlet extractors, complete assemblies
Supplier: SciLabware
Certificates
About this item
PYREX®, borosilicate glass.
- Complete glass assembly
- Low solvent volumes required for extraction of soluble components in solids
- Range of sizes of extractor to meet different sample sizes
* 75 mm diameter extractor flange.
Delivery: Assembly consists of round bottom flask, Soxhlet extractor and Allihn condenser.
Frequently Bought Together