Blood lancets, BD Microtainer® contact-activated
Supplier: BD Medical
Certificates
About this item
BD Microtainer® contact-activated blood lancet is a safety-engineered device used for fingerstick collection of blood.
- Enhanced user comfort
- Activates on contact
- Simplifies the procedure
Caution: Fingersticks should not be performed on children less than one year of age according to CLSI guidelines.
Frequently Bought Together