2,4-Dichlorobenzaldehyde 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:874-42-0
:
Formula:C₇H₄Cl₂O
MDL Number:MFCD00003305
Boiling Pt:233
Molecular Weight:175.01 g/mol
Melting Pt:69…73 °C
Flash Pt:135 °C
Storage Temperature:Ambient
EINECS:212-861-1
UN:3261
ADR:8 III
:GHS05
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Light yellow to beige Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:68 to 73 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together