Methyl heptadecanoate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Heptadecanoic acid methyll ester, Methyl Margarate, Margaric acid methyll ester
CAS Number:1731-92-6
:
Formula:C₁₈H₃₆O₂
MDL Number:MFCD00009001
Molecular Weight:284.48 g/mol
:M-Clarity™ quality level = MQ200
EINECS:217-055-3
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together