2-Hydroxyethyl methacrylate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: HEMA, GMA, Methacrylic acid 2-hydroxyethyl ester, 1,2-Ethanediol mono(2-methylpropenoate), Glycol methacrylate
CAS Number:868-77-9
:
Formula:C₆H₁₀O₃
MDL Number:MFCD00002863
Molecular Weight:130.14 g/mol
:M-Clarity™ quality level = MQ200
EINECS:212-782-2
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together