Triisopropanolamine, Sigma-Aldrich®
: Merck
: 1,1',1''-Nitrilotripropan-2-ol, 1,1',1"-Nitrilotri-2-propanol, Tris(2-hydroxypropyl)amine
:122-20-3
:
:C₉H₂₁NO₃
:MFCD00004533
:191.27 g/mol
:M-Clarity™ quality level = MQ200
:204-528-4
:GHS07
:Ambient
- Environmentally Preferable:
- Solubility:(20 °C) soluble
- Melting Point:50 °C
- Molar Mass:191.27 g/mol
- Boiling Point:305 °C (1013 hPa)
- Vapor Pressure:1 hPa (100 °C)
- Flash Point:160 °C
- Explosion Limit:0.8 - 5.8 %(V)
- Density:1.01 g/cm3 (50 °C)
- pH:10.8 (100 g/l, H2O, 20 °C)
- Ignition Point:275 °C
Frequently Bought Together