Order Entry
Austria
ContactUsLinkComponent
2-Methylbutane ≥99%, GPR RECTAPUR®
2-Methylbutane ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
2-Methylbutane ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Boiling point:
    27.0 - 28.5 °C
  • Density (20/4):
    0.600 - 0.640
  • Refractive index (20°C):
    1.353 - 1.355
  • Evaporation residue:
    Max. 50 ppm

Specification Test Results

Frequently Bought Together