2,2-Dimethylbutane
Catalog # EHERCA12726100
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
- Environmentally Preferable:
Specifications
About this item
Synonyms: Neohexane
:
:86.18 g/mol
EINECS:200-906-8
UN:1208
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:75-83-2
Formula:C₆H₁₄
MDL Number:MFCD00009321
Boiling Pt:49.7
Melting Pt:–100 °C
Density:0.649 g/cm³ (20 °C)
ADR:3 II