1,2-Dichloropropane 98%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Propylene dichloride
CAS Number:78-87-5
:
Formula:ClH₂CCH₂CH₂Cl
MDL Number:MFCD00000868
Boiling Pt:95…96 °C (760 mmHg)
Melting Pt:–100 °C
Density:1.156 g/cm³
Molecular Weight:112.99 g/mol
Flash Pt:15 °C
Storage Temperature:Ambient
UN:1279
ADR:3 II
Merck Index:12,08039
EINECS:201-152-2
:GHS02
:GHS08
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4380 to 1.4400 (20°C, 589 nm)
- GC:≥97.5 %
- Ash:≤4.5 % (825°C)
- pH:6.5 to 7 (10 % at 20°C)
Specification Test Results
Frequently Bought Together