2-Methyl-4(5)-nitroimidazole 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Methyl-4-nitro-1H-imidazole, 2-Methyl-5-nitroimidazole, 2-Methyl-5-nitro-1H-imidazole, 2-Methyl-4-nitroimidazole
CAS Number:88054-22-2
Formula:C₄H₅N₃O
MDL Number:MFCD00005191
Molecular Weight:127.10 g/mol
Melting Pt:252…254 °C
EINECS:211-790-3
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to beige Powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥98.5 %
- Sodium dihydrogenphosphate:≤2.5 %
Specification Test Results
Frequently Bought Together