Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-3-chloro-5-(trifluoromethyl)pyridine
2-Bromo-3-chloro-5-(trifluoromethyl)pyridine
2-Bromo-3-chloro-5-(trifluoromethyl)pyridine
MAYBBTB09453DA
:
Thermo Scientific
:
2-Bromo-3-chloro-5-(trifluoromethyl)pyridine
MAYBBTB09453DA
:
Thermo Scientific
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
Environmentally Preferable:
:
:
260.44 g/mol
:
75806-84-7
:
C₆H₂BrClF₃N
:
MFCD00153072