Sodium carbonate decahydrate 99+%, extra pure
: Soda
:6132-02-1
:
:Na₂CO₃·10H₂O
:MFCD00149178
:1.46 g/cm³
:286.14 g/mol
:34 °C
:207-838-8
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:White Crystals
- Titration with HCl:≥99.0 %
- Chloride (Cl):≤10 ppm
- Sulfate (SO4):≤30 ppm
- Aluminium (Al):≤10 ppm
- Iron (Fe):≤4 ppm
- Magnesium (Mg):≤100 ppm
- Potassium (K):≤50 ppm
- Silicon dioxide (SiO2):≤30 ppm
Frequently Bought Together