Order Entry
Austria
ContactUsLinkComponent
Sodium hydroxide 50% (w/w) in water, extra pure
Sodium hydroxide 50% (w/w) in water, extra pure
Certificates
undefined
Sodium hydroxide 50% (w/w) in water, extra pure
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Viscous solution
  • Heavy metals:
    ≤10 ppm
  • Density:
    (20°C) 1.510 to 1.540 kg/l
  • Titration with HCl:
    49.0 to 51.0 %
  • Sodium carbonate (Na2CO3):
    ≤0.2 % (Na2CO3)
  • Sodium sulfate (Na2SO4):
    ≤100 ppm (Na2SO4)
  • Sodium chloride (NaCl):
    ≤200 ppm (NaCl)

Specification Test Results

Related Information

Frequently Bought Together