Order Entry
Austria
ContactUsLinkComponent
Sodium chloride 180 g/l in aqueous solution
Sodium chloride 180 g/l in aqueous solution
Catalog # 230386M
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium chloride 180 g/l in aqueous solution
Catalog # 230386M
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (W/V):
    17.9 - 18.1 %
  • pH (20°C):
    5.5 - 8.5

Specification Test Results

About this item

Synonyms: Saline, NaCl, Sea salt, Halite
 :
 :58.44 g/mol
Boiling Pt:1413
Melting Pt:801
Density:1.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477
Merck Index:14,08599