ContactUsLinkComponent
Homogeniser disposable pestles and tubes, polypropene, KIMBLE®
Catalog # SCERSP749540-0000
Supplier:  DWK Life Sciences
undefined
Homogeniser disposable pestles and tubes, polypropene, KIMBLE®
Catalog # SCERSP749540-0000
Supplier:  DWK Life Sciences
Specifications
  • Description:
    Motor, with adapter
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Stainless steel cordless drive unit. Pressure is 14.7 PSI.
  • Resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
  • Operates at 2000 to 3000 min⁻¹ with fresh batteries and no load
Delivery: Supplied with 2× AA batteries.