Stools, mobile, metal
Manufactured from strong powder coated steel and mounted on three castors allowing complete mobility. When in use the castors retract, providing a secure platform.
- Non slip rubber mats, lead- and cadmium-free lacquer
- TÜV GS approved, supports up to 150 kg
- Snap-lock assembly, no tools required
EN 14183 (DIN 4569)
: EN 14183 (DIN 4569).
Frequently Bought Together