Bottle top vacuum filtration systems, Nalgene®
PS, reusable bottle top vacuum filtration funnels suitable for analytical or sterile filtration, with vacuum or pressure.
- With graduation and screw thread
- Removable membrane support plate
- PP tubing adapter
513-3203: For neck size 33 mm
516-0578 and 516-0577: For neck size 45 mm
PP tubing adapter
513-3203: for neck size 33 mm
516-0578: for neck size 45 mm
: The filter is not included and has to be ordered separately. It should have a diameter of 47 - 50 mm.
- Catalog No:
- 516-0577
- 513-3203
- 516-0578
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Frequently Bought Together