About this item
Vivaspin® 20 centrifugal concentrators offer increased volume flexibility and performance to the Vivaspin® potfolio. Vivaspin® 20 handles 5 to 20 ml in swing bucket rotors and 14 ml in fixed angle rotors accepting 50 ml centrifuge tubes. Vivaspin® 20 features twin vertical membranes for unparalleled filtration speed and >100-fold concentration. Volume graduation allows monitoring the concentration process and the modified dead stop pocket simplifies pipetting of the final concentrate.
- High retentate recovery >90%
- Vertical twin membrane for ultra-fast concentration
- Safe sample concentration due to dead-stop volume
- Convenient concentrate recovery by pipetting
- Volume graduation for easy reading/monitoring
For higher process flexibility, Vivaspin® 20 is available with unique accessories and operating methods. For one-step desalting following concentration, a diafiltration cup filled with buffer can be placed on the concentrator and spun another time to achieve 98% salt removal, instead of two spins to achieve the same result with the traditional refill and re-spin procedure. When an appropriate centrifuge is unavailable, or for single sample processing, for gas pressure filtration, Vivaspin® 20 can be filled with up to 15 ml and then pressurised for bench top concentration. For even faster processing, gas pressure filtration can be combined with centrifugation. 'Pressure-fugation' is particularly useful for difficult or viscous samples such as serum, when using a low process temperature, or generally when minimum process time is essential.
Specifications
- Catalog No:
- 512-3785
- 512-2863
- 512-4022
- 512-3784
- 512-4021
- 512-2862
- 512-3783
- 512-4020
- 512-2864
- 512-4018
- 512-4024
- 512-2865
- 512-3788
- 512-4019
- 512-3787
- 512-3786
- Filtration area:
- 6 cm²
- 6 cm²
- 6 cm²
- 6 cm²
- 6 cm²
- 6 cm²
- 6 cm²
- 6 cm²
- 6 cm²
- 6 cm²
- 6 cm²
- 6 cm²
- 6 cm²
- 6 cm²
- 6 cm²
- 6 cm²
- Max. RCF:
- 8000 ×g
- 8000 ×g
- 8000 ×g
- 8000 ×g
- 8000 ×g
- 8000 ×g
- 8000 ×g
- 8000 ×g
- 8000 ×g
- 8000 ×g
- 8000 ×g
- 8000 ×g
- 8000 ×g
- 8000 ×g
- 8000 ×g
- 8000 ×g
- Dead volume:
- <20 µl
- <20 µl
- <20 µl
- <20 µl
- <20 µl
- <20 µl
- <20 µl
- <20 µl
- <20 µl
- <20 µl
- <20 µl
- <20 µl
- <20 µl
- <20 µl
- <20 µl
- <20 µl
- Ø×H:
- 30×116 mm
- 30×116 mm
- 30×116 mm
- 30×116 mm
- 30×116 mm
- 30×116 mm
- 30×116 mm
- 30×116 mm
- 30×116 mm
- 30×116 mm
- 30×116 mm
- 30×116 mm
- 30×116 mm
- 30×116 mm
- 30×116 mm
- 30×116 mm
Specifications
Related Information
Certificates
Search by Lot NumberAppl_Ultrafiltration_work_flow_SLU4000-e Appl_Urine_Protein_Concentration_SL-4069-e Appl_Vivaspin-20_Diafiltration-cups_SL-4076-e Appl_Vivaspin500_SL-4066-e Appl_Vivaspin_Desalting_SLL4002-e
Centrifugal Filtration Units, Vivaspin® Sartorius
Application Notes
SARTVS2001_TDS_EN SARTVS2002_TDS_EN