pGFP-C-shLenti shRNA Vector
Catalog # ORIGTR30023
Supplier: OriGene
New Product
CAS Number:
Specifications
- Pk:5 µG
- Description:pGFP-C-shLenti shRNA Vector
- Environmentally Preferable:
- Storage temperature:Store the vector DNA at –20 °C.
- Vector details:HuSH shRNA GFP Lenti cloning vector
Specifications
About this item
pGFP-C-shLenti vector is a third generation Lentivector which requires the viral components carried in other vectors to produce viral particles. There are three major functional elements within the 5LTR and 3LTR: a shRNA expression cassette driven by an U6 promoter, a puromycin resistant gene driven by a SV40 promoter and a tGFP gene driven by a CMV promoter. All of them can be packaged to viral particles and transduced to many cell lines. The bacterial selection marker for the vector is Chloramphenicol.
- U6_F Primer can be used to sequence the target sequence after cloning
- Components: 1 vial of 5 ug lyophilized pGFP-C-shLenti shRNA cloning vector
- Stability: Vector DNA is stable for at least one year from date of receipt when stored at –20 °C