Order Entry
Austria
ContactUsLinkComponent
Anti-Von Hippel Lindau/VHL Mouse Monoclonal Antibody [clone: ABT-PVHL]
Anti-Von Hippel Lindau/VHL Mouse Monoclonal Antibody [clone: ABT-PVHL]
Supplier:  ANTIBODIES.COM
New Product
Certificates
undefined
Anti-Von Hippel Lindau/VHL Mouse Monoclonal Antibody [clone: ABT-PVHL]
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Catalog No:
  • ANTIA100015-50
  • ANTIA100015-100
  • Antigen symbol:
  • Von Hippel Lindau/VHL
  • Von Hippel Lindau/VHL
  • Antigen name:
  • Elongin binding protein
  • Elongin binding protein
  • Antigen synonyms:
  • VHL 1|Protein G7|VHLH|G7 protein|pVHL|VHL1|HRCA 1|VHL_HUMAN|HRCA1
  • VHL 1|Protein G7|VHLH|G7 protein|pVHL|VHL1|HRCA 1|VHL_HUMAN|HRCA1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • ABT-PVHL
  • ABT-PVHL
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • UniprotID# P40337
  • UniprotID# P40337
  • Isotype:
  • IgG
  • IgG
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic peptide derived from human pVHL
  • Synthetic peptide derived from human pVHL
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Immunogen affinity purified
  • Immunogen affinity purified
  • Storage buffer:
  • Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
  • Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
  • Shipping temperature:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C
  • Tested applications:
  • IHC
  • IHC

Specifications

Frequently Bought Together