Anti-HCG alpha Mouse Monoclonal Antibody [clone: hCGa/7874]
Catalog # ANTIA316448-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Anterior pituitary glycoprotein hormones common subunit alpha
- Antigen symbol:HCG alpha
- Clonality:Monoclonal
- Clone:hCGa/7874
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P01215
- Antigen synonyms:Follicle-stimulating hormone alpha|Chorionic Gonadotrophin Alpha Subunit|CG alpha|Chorionic Gonadotrophin Alpha|Choriogonadotropin alpha chain|CG-alpha|cga|Chorionic gonadotrophin subunit alpha|chorionic gonadotropin, alpha polypeptide
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:13 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-116) of human hCG alpha protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [hCGa/7874] antibody to HCG alpha for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: HCG alpha
Clonality: Monoclonal
Clone: hCGa/7874
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human