Butyl (±)-lactate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Butyl 2-hydroxypropanoate, n-Butyl lactate, Butyl (±)-2-hydroxypropionate, (±)-Butyl lactate
CAS Number:138-22-7
:
Formula:C₇H₁₄O₃
MDL Number:MFCD00004519
Boiling Pt:170
Melting Pt:–28 °C
Density:0.9832 g/cm³ (20 °C)
Molecular Weight:146.19 g/mol
:M-Clarity™ quality level = MQ200
EINECS:205-316-4
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together