L(-)-Sorbose, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: L-(-)-Sorbose, (3S,4R,5S)-1,3,4,5,6-Pentahydroxyhexan-2-one
CAS Number:87-79-6
:
Formula:C₆H₁₂O₆
MDL Number:MFCD00151097
Boiling Pt:380
Molecular Weight:180.16 g/mol
Melting Pt:158…160 °C
Merck Index:12,08874
Storage Temperature:Ambient
EINECS:201-771-8
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together