Order Entry
Austria
ContactUsLinkComponent
Phenylpropiolic acid
Phenylpropiolic acid
Catalog # MOLE35389201-25G
Supplier:  Molekula
CAS Number:  
Phenylpropiolic acid
Catalog # MOLE35389201-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Phenylacetylenecarboxylic acid, Phenylpropynoic acid, 3-Phenylpropiolic acid
 :
 :146.15 g/mol
EINECS:211-285-8
 :GHS07
CAS Number:637-44-5
Formula:C₉H₆O₂
MDL Number:MFCD00004361
Boiling Pt:294
Melting Pt:135…138 °C
Density:1.28 g/cm³ (20 °C)