5-(Hydroxymethyl)-2-furaldehyde for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 5-Hydroxymethyl-2-furancarbaldehyde, 5-(hydroxymethyl)furan-2-carbaldehyde, 5-(Hydroxymethyl)furfural, 5-Hydroxymethylfurfural
CAS Number:67-47-0
:
Formula:C₆H₆O₃
MDL Number:MFCD00003234
Boiling Pt:292
Melting Pt:27…30 °C
Density:1.24 g/cm³ (20 °C)
Molecular Weight:126.11 g/mol
Flash Pt:79
Merck Index:13,04857
:M-Clarity™ quality level = MQ200
EINECS:200-654-9
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:27 - 30 °C
- Molar Mass:126.11 g/mol
- Boiling Point:154 - 155 °C (16 hPa)
- Flash Point:79 °C
- Refractive Index:1.5627 (20 °C, 589 nm)
- Synonyms:5-(Hydroxymethyl)furfurol
- Hill Formula:C6H6O3
Specification Test Results
Related Information
Frequently Bought Together