4-Propylcyclohexanone for synthesis, Sigma-Aldrich®
: Merck
:40649-36-3
:
:C₉H₁₆O
:MFCD00058690
:219
:0.91 g/cm³ (20 °C)
:140.23 g/mol
:M-Clarity™ quality level = MQ200
:406-810-4
:GHS07
- Environmentally Preferable:
- Solubility:1.96 g/l (20 °C)
- Molar Mass:140.22 g/mol
- Boiling Point:219 °C (1013 hPa)
- Vapor Pressure:0.23 hPa (20 °C)
- Flash Point:90 °C
- Density:0.91 g/cm3 (20 °C)
- pH:6.0 (2 g/l, H2O, 20 °C)
- Ignition Point:320 °C
Frequently Bought Together