Order Entry
Austria
ContactUsLinkComponent
cis-1,2-Cyclopentanediol 98%
cis-1,2-Cyclopentanediol 98%
Certificates
undefined
cis-1,2-Cyclopentanediol 98%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid after melting
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4770 to 1.4790 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together