Tris(trimethylsilyl)silane 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1,1,1,3,3,3-Hexamethyl-2-(trimethylsilyl)trisilane, TTMSS
CAS Number:1873-77-4
:
Formula:C₉H₂₈Si₄
MDL Number:MFCD00077893
Boiling Pt:82…84 °C (12 mmHg)
Density:0.8 g/cm³
Molecular Weight:248.66 g/mol
Flash Pt:55 °C
UN:1993
ADR:3 III
Merck Index:12,09886
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4890 to 1.4910 (20°C, 589 nm)
- GC:≥95 %
Specification Test Results
Frequently Bought Together