Order Entry
Austria
ContactUsLinkComponent
 
3-Phenylpropanal 95%
 
undefined
3-Phenylpropanal 95%

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5200 to 1.5300 (20°C, 589 nm)
  • GC:
    ≥94.0 %

 

 

Frequently Bought Together