Sodium perborate tetrahydrate 97%
: Sodium peroxometaborate tetrahydrate
:10486-00-7
:NaH₂BO₄·4H₂O
:MFCD00149231
:171.87 g/mol
:60
:239-172-9
:GHS07
:GHS05
:GHS08
:Ambient
- Environmentally Preferable:
- Appearance:White Crystalline powder
- pH:10.1 to 10.6 (1 % solution)
- Titration with KMnO4:≥96.0 %
Frequently Bought Together