Methyl bromoacetate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Bromoacetic methyl ester
CAS Number:96-32-2
:
Formula:BrCH₂COOCH₃
MDL Number:MFCD00000189
Boiling Pt:145
Melting Pt:–50 °C
Density:1.61 g/cm³
Molecular Weight:152.98 g/mol
Flash Pt:62 °C
Storage Temperature:Refrigerator
EINECS:202-499-2
UN:2643
ADR:6.1 II
:GHS05
:GHS06
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4570 to 1.4590 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together