Order Entry
Austria
ContactUsLinkComponent
 
Levulinic acid 98+%
 
undefined
Levulinic acid 98+%

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Colorless to white to yellow to orange Liquid after melting or solid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4390 to 1.4430 (20°C, 589 nm)
  • Titration with NaOH:
    ≥98.0 %

 

 

Frequently Bought Together