Citric acid, anhydrous ≥99.5%, granular FCC
Catalog # SPCMC1281-500GM
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Poly bottle
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (C₆H₈O₇ ;Anhyd.,Basis):99.5 - 100.5%
- Lead (Pb):max. 0.5 mg/kg
- Oxalate (C₂O₄):To Pass Test
- Readily Carbonizable Substances:To Pass Test
- Residue On Ignition:max. 0.05 %
Specification Test Results
About this item
Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
:
:192.13 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:77-92-9
Formula:HOC(COOH)(CH₂COOH)₂
MDL Number:MFCD00011669
Boiling Pt:310
Merck Index:13,02350