- Antibody type:Primary
- Antigen name:BAGE4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Epitope:18-39/39
- Form:Liquid
- Gene ID:3624
- Antigen synonyms:BAGE4_HUMAN|Cancer/testis antigen 2.4|MGC119851|CT2.4|BAGE4|B melanoma antigen family, member 4|MLL3P|MGC119852|MGC119853|B melanoma antigen 4
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human BAGE4
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Members of the BAGE gene family encode antigens that are recognised by cytotoxic T lymphocytes and are also known as CT (cancer/testis) antigens. Generated by juxtacentromeric shuffling of the MLL3 gene, the ancestral BAGE gene was expanded by acrocentric exchanges and/or juxtacentromeric movements. BAGE, also known as B melanoma antigen 1 or cancer/testis antigen 2.1, is a 43 amino acid member of the BAGE family. Generally, BAGE proteins are not expressed in normal tissues, except in testis, but are expressed highly in melanomas, bladder carcinomas, head and neck squamous cell carcinomas, and lung and breast cancer carcinomas. BAGE proteins are not expressed in renal, colorectal and prostatic carcinomas, leukemias and lymphomas.
Type: Primary
Antigen: BAGE4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 18-39/39
Host: Rabbit
Isotype: IgG
Reactivity: Human